Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

61724

Millipore

Liver Broth

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171607
NACRES:
NA.74

sterility

non-sterile

Quality Level

form

solid

shelf life

limited shelf life, expiry date on the label

composition

disodium hydrogen phosphate, 2 g/L
D(+)-glucose, 10 g/L
liver powder, 50 g/L

final pH

7.0±0.2 (25 °C)

application(s)

food and beverages
microbiology

suitability

anaerobic bacteria
nonselective for Clostridium spp.

Application

For preliminary testing and for the enrichment of Clostridia and other anaerobes from meat, food and other material according to Kelch.

Other Notes

Bacteriological diagnosis of Vibrio fetus infection in man[1]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F. Kelch
Archiv Fur Lebensmittelhygiene, 6, 263-263 (1955)
U Ullmann
Zentralblatt fur Bakteriologie, Parasitenkunde, Infektionskrankheiten und Hygiene. Erste Abteilung Originale. Reihe A: Medizinische Mikrobiologie und Parasitologie, 230(4), 480-491 (1975-01-01)
A synopsis of the present classification of Vibrio fetus is given (tab. 1, 2). Besides the presentation of the importance of these bacteria in veterinary medicine, 97 human infections have been published up to now. The geographic, age and sex

Articles

Microbiological assessment of bacteria found in spices, a potent source for food spoilage and pathogens.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service