Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

61600

Sigma-Aldrich

Laurolactam

puriss., ≥99.0% (N)

Synonym(s):

12-Aminododecanoic acid lactam, Aza-2-cyclotridecanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H23NO
CAS Number:
Molecular Weight:
197.32
Beilstein/REAXYS Number:
122031
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

puriss.

assay

≥99.0% (N)

form

solid

mp

149-153 °C

SMILES string

O=C1CCCCCCCCCCCN1

InChI

1S/C12H23NO/c14-12-10-8-6-4-2-1-3-5-7-9-11-13-12/h1-11H2,(H,13,14)

InChI key

JHWNWJKBPDFINM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Ganry et al.
International journal of oral and maxillofacial surgery, 46(8), 946-957 (2017-04-24)
Very few surgical teams currently use totally independent and free solutions to perform three-dimensional (3D) surgical modelling for osseous free flaps in reconstructive surgery. This study assessed the precision and technical reproducibility of a 3D surgical modelling protocol using free
Magdalena Kwiatkowska et al.
Polymers, 12(2) (2020-02-08)
In this work the preparation of polyamide 12 (PA12) based composites reinforced with pristine and surface-modified carbon nanotubes is reported. A qualitative and quantitative evaluation of multi-walled carbon nanotube functionalization with oxygen containing reactive groups achieved by different procedures of
Coraline Simon et al.
Toxicology in vitro : an international journal published in association with BIBRA, 37, 121-133 (2016-10-19)
Endocrine activity of 65 compounds migrating from polycarbonate replacement plastic baby bottles was assessed using in vitro cell based assays (reporter gene assays) involving 7 nuclear receptors, i.e. human steroid hormones receptors (oestrogen, androgen, progesterone and glucocorticoid receptors), human thyroid
Matthias Onghena et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 31(12), 2090-2102 (2014-11-20)
In 2011, the European Union prohibited the production of polycarbonate (PC) baby bottles due to the toxic effects of the PC monomer bisphenol-A. Therefore, baby bottles made of alternative materials, e.g. polypropylene (PP) or polyethersulphone (PES), are currently marketed. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service