Skip to Content
MilliporeSigma

61430

Laminarin from Laminaria digitata

Synonym(s):

Laminaran

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352201
EC Number:
232-712-4
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


solubility

H2O: 20 mg/mL, clear, faintly yellow

InChI

1S/C18H32O16/c19-1-4-7(22)10(25)11(26)17(31-4)34-15-9(24)6(3-21)32-18(13(15)28)33-14-8(23)5(2-20)30-16(29)12(14)27/h4-29H,1-3H2/t4-,5-,6-,7-,8-,9-,10+,11-,12-,13-,14+,15+,16?,17?,18?/m1/s1

InChI key

DBTMGCOVALSLOR-VPNXCSTESA-N

Other Notes

Characterization of β-glucanases from S. cerevisiae




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Saccharomyces cerevisiae secretes 2 exo-beta-glucanases.
A Sánchez et al.
FEBS letters, 138(2), 209-212 (1982-02-22)