Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

61338

Supelco

Norfentanyl monohydrate

analytical standard

Synonym(s):

4-(N-Phenyl-N-propionylamino)piperidine, N-4-Piperidylpropionanilide, N-Phenyl-N-4-piperidinylpropanamide, N-Phenyl-N-4-piperidinylpropionamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H20N2O· H2O
CAS Number:
Molecular Weight:
250.34
Beilstein/REAXYS Number:
480031
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical

format

neat

SMILES string

CCC(N(C1=CC=CC=C1)C2CCNCC2)=O.O

InChI

1S/C14H20N2O.H2O/c1-2-14(17)16(12-6-4-3-5-7-12)13-8-10-15-11-9-13;/h3-7,13,15H,2,8-11H2,1H3;1H2

InChI key

MHZWGPZXICYPPJ-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service