Sign In to View Organizational & Contract Pricing.
Select a Size
About This Item
Empirical Formula (Hill Notation):
C12H24O11 · H2O
CAS Number:
Molecular Weight:
362.33
EC Number:
209-566-5
UNSPSC Code:
12352201
PubChem Substance ID:
Beilstein/REAXYS Number:
4218679
MDL number:
assay
≥99.0% (HPLC)
optical activity
[α]20/D +13.5±1°, c = 1% in H2O
mp
95-98 °C (lit.)
SMILES string
[H]O[H].OC[C@H](O)[C@@H](O)[C@H](O[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O)[C@H](O)CO
InChI
1S/C12H24O11.H2O/c13-1-4(16)7(18)11(5(17)2-14)23-12-10(21)9(20)8(19)6(3-15)22-12;/h4-21H,1-3H2;1H2/t4-,5+,6+,7+,8-,9-,10+,11+,12-;/m0./s1
InChI key
LXMBXZRLTPSWCR-XBLONOLSSA-N
Biochem/physiol Actions
Sugar alcohol sweet tastant detectable by humans.
Storage Class
13 - Non Combustible Solids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service