Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

61313

Supelco

L-α-Hydroxyglutaric acid disodium salt

analytical standard

Synonym(s):

(S)-2-Hydroxypentanedioic acid disodium salt, Disodium (S)-2-hydroxyglutarate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H6O5Na2
CAS Number:
Molecular Weight:
192.08
Beilstein/REAXYS Number:
5318042
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

assay

≥98.0% (GC)

optical activity

[α]/D -8.5±1.5°, c = 1 in 0.1 M NaOH

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

[O-]C(CC[C@@H](C([O-])=O)O)=O.[Na+].[Na+]

InChI

1S/C5H8O5.2Na/c6-3(5(9)10)1-2-4(7)8;;/h3,6H,1-2H2,(H,7,8)(H,9,10);;/q;2*+1/p-2/t3-;;/m0../s1

InChI key

DZHFTEDSQFPDPP-QTNFYWBSSA-L

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

L-alpha-Hydroxyglutaric acid accumulates as a result of a rare defect in L-2-HG dehydogenase, leading to the metabolic disorder L-2-hydroxyglutaric aciduria (L-2HGA). L-2HGA is associated with neuronal defects, leukodystrophy and linked to an increased risk of brain tumors.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service