Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

61227

Supelco

L-Serine hydrochloride solution

100 mM amino acid in 0.1 M HCl, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H7NO3 · HCl
CAS Number:
Molecular Weight:
141.55
Beilstein/REAXYS Number:
3984536
MDL number:
UNSPSC Code:
85151701
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.24
Product 61227 is not currently sold in your country. Contact Technical Service

grade

analytical standard

Quality Level

form

liquid

analyte chemical class(es)

amino acids, peptides, proteins

concentration

100 mM amino acid in 0.1 M HCl

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

colorless

application(s)

food and beverages

format

single component solution

storage temp.

2-8°C

SMILES string

Cl.N[C@@H](CO)C(O)=O

InChI

1S/C3H7NO3.ClH/c4-2(1-5)3(6)7;/h2,5H,1,4H2,(H,6,7);1H/t2-;/m0./s1

InChI key

STMOVTSFWYRCOB-DKWTVANSSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service