Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

61221

Supelco

Castalagin

analytical standard

Synonym(s):

(33β)-Vescalagin, NSC 297535

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H26O26
CAS Number:
Molecular Weight:
934.63
UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

impurities

≤15.0% water

application(s)

food and beverages

format

neat

shipped in

wet ice

storage temp.

−20°C

SMILES string

O=C(C1=CC(O)=C(O)C(O)=C1C2=C(C(OC[C@@]3([H])O4)=O)C=C(O)C(O)=C2O)O[C@@]3([H])[C@H](O5)[C@@]([C@H]6O)([H])OC(C7=C6C(O)=C(O)C(O)=C7C8=C(O)C(O)=C(O)C(C9=C(O)C(O)=C(O)C=C9C4=O)=C8C5=O)=O

InChI

1S/C41H26O26/c42-8-1-5-12(24(48)21(8)45)13-6(2-9(43)22(46)25(13)49)39(60)65-34-11(4-63-37(5)58)64-38(59)7-3-10(44)23(47)26(50)14(7)15-18-16(28(52)32(56)27(15)51)17-19-20(30(54)33(57)29(17)53)31(55)35(66-41(19)62)36(34)67-40(18)61/h1-3,11,31,34-36,42-57H,4H2

InChI key

UDYKDZHZAKSYCO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Castalagin is a polyphenol that is mainly present in oak and chestnut wood.[1] It belongs to the class of hydrolyzable tannins or ellagitannins that is reported to exhibit anti-inflammatory, anticancer, antioxidant, and antibacterial properties. The anti-herpes virus activity of the compound is also investigated.[2][3][4]

Application

Castalagin may be used as an analytical standard for the determination of the analyte in oak species,[5] and various parts of white strawberry species[6] by chromatography techniques.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ellagitannins and Flavano-Ellagitannins: Red Wines Tendency in Different Areas, Barrel Origin and Ageing Time in Barrel and Bottle
Rasines-Perea Z, et al.
Food Chemistry, 9(8), 316-316 (2019)
Analysis of oak tannins by liquid chromatography-electrospray ionisation mass spectrometry
Mammela P, et al.
Journal of Chromatography A, 891(1), 75-83 (2000)
Castalagin exerts inhibitory effects on osteoclastogenesis through blocking a broad range of signaling pathways with low cytotoxicity
Iwatake M, et al.
Phytotherapy Research, 29(6), 917-924 (2015)
Determination of phenolic composition and antioxidant activity in fruits, rhizomes and leaves of the white strawberry (Fragaria chiloensis spp. chiloensis form chiloensis) using HPLC-DAD-ESI-MS and free radical quenching techniques
Simirgiotis MJ and Schmeda-Hirschmann G
Journal of food composition and analysis, 23(6), 545-553 (2010)
Three-dimensional analysis of combination effect of ellagitannins and acyclovir on herpes simplex virus types 1 and 2
Vilhelmova N, et al.
Antiviral Research, 89(2), 174-181 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service