Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

611921

Supelco

Hexamethyldisiloxane solution

25% in benzene-d6 (99.6 atom % D), NMR tube size 5 mm × 8 in.

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H18OSi2
Molecular Weight:
162.38
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

concentration

25% in benzene-d6 (99.6 atom % D)

technique(s)

NMR: suitable

NMR tube size

5 mm × 8 in.

suitability

suitable for NMR (reference standard)

format

single component solution

SMILES string

C[Si](C)(C)O[Si](C)(C)C

InChI

1S/C6H18OSi2/c1-8(2,3)7-9(4,5)6/h1-6H3

InChI key

UQEAIHBTYFGYIE-UHFFFAOYSA-N

Features and Benefits

29Si sensitivity

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Carc. 1A - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 1B - Skin Irrit. 2 - STOT RE 1

target_organs

Blood

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

12.2 °F - closed cup

flash_point_c

-11 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service