Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

610658

Supelco

Methaqualone-d7 solution

1 mg/mL in methanol, drug standard, 98 atom % D

Synonym(s):

Methaqualone-tolyl-d7 solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16D7H7N2O
CAS Number:
Molecular Weight:
257.34
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

drug standard

isotopic purity

98 atom % D

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IIC (Portugal)

availability

not available in Japan

concentration

1 mg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

mass shift

M+7

storage temp.

−20°C

SMILES string

[2H]c1c([2H])c([2H])c(c(c1[2H])N2C(C)=Nc3ccccc3C2=O)C([2H])([2H])[2H]

InChI

1S/C16H14N2O/c1-11-7-3-6-10-15(11)18-12(2)17-14-9-5-4-8-13(14)16(18)19/h3-10H,1-2H3/i1D3,3D,6D,7D,10D

InChI key

JEYCTXHKTXCGPB-ZMMMWQAQSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service