61001BC
TRANSIA® AG Salmonella Kit
suitable for ELISA testing in Environmental, and Food and Beverages
About This Item
Recommended Products
Product Name
TRANSIA® AG Salmonella, BioControl, ELISA test for detection of Salmonella in food and environmental samples
agency
AOAC® (OMA)
form
liquid
shelf life
3‑9 mo.
packaging
pkg of 504
pkg of 96
manufacturer/tradename
BioControl Systems 61001-504
BioControl Systems 61001-96
TRANSIA®
technique(s)
ELISA: suitable
input
food(s)
application(s)
environmental
food and beverages
pathogen testing
storage temp.
2-8°C
suitability
Salmonella spp.
General description
Application
Features and Benefits
- Approved for use with all foods.
- Provides next day results for processed food while raw foods require one additional day.
- Results can be read visually or with an automated microplate reader.
- Ready to use reagents and breakaway wells allow you to use only the number of tests required for your samples, helping to reduce waste and keep testing costs low.
Components
- Test Wells
- Reagent 1 - Extraction Reagent
- Reagent 2 - Wash Solution Concentrate
- Reagent 3 - Positive Control
- Reagent 4 - Conjugate
- Reagent 5 - Substrate
- Reagent 6 - Stop Solution
- Color Standard Card
Testing Method
Legal Information
Kit Components Also Available Separately
- Extraction Reagent, 12.5 mL, 96 tests
- Wash Solution, 55 mL, 96 tests
- Positive Control, 11 mL, 96 tests
- Conjugate Solution 16 mL, 96 tests
- Substrate, 34005-504, 57 mL, 96 tests
- Stop Solution, 57 mL, 96 tests
- Antibody wells, 96 tests
signalword
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2 - Met. Corr. 1 - Repr. 1B - Skin Irrit. 2 - Skin Sens. 1
supp_hazards
Storage Class
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Updates on ISO standards in food microbiology by expert Barbara Gerten from MilliporeSigma
Related Content
Food microbiology testing is a key part of the food safety and quality management. Learn more about the wide range of tests covering pathogens and spoilage organisms.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service