Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

60758

Supelco

C18-Reversed phase silica gel

for column chromatography, not endcapped

Synonym(s):

C18-reversed phase silica gel, Reversed phase silica gel C18

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
MDL number:
UNSPSC Code:
12352200

grade

for column chromatography

form

gel form

quality

not endcapped

technique(s)

LPLC: suitable

surface coverage

surface coverage (C) 8‑10% 2.0 μmol/m2

matrix

Silica

matrix active group

C18 phase

particle size

230-400 mesh
40-63 μm

pore size

90 Å pore size

suitability

suitable for column chromatography

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

Application

for reversed phase chromatography; designed for special tuning of selectivity

Storage Class

11 - Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service