Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

60641

Millipore

KF-Streptococcus Agar

suitable for microbiology, NutriSelect® Plus

Synonym(s):

Kenner Fecal Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.74

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 20 g/L
bromocresol purple, 0.015 g/L
lactose, 1 g/L
maltose, 20 g/L
proteose peptone, 10 g/L
sodium azide, 0.4 g/L
sodium chloride, 5 g/L
sodium glycerophosphate, 10 g/L
yeast extract, 10 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

7.2±0.2 (25 °C)

application(s)

bioburden testing
environmental
food and beverages
water monitoring

microbiology

suitability

selective and differential for Enterococcus spp.
selective and differential for Streptococcus spp.

Application

For the detection and enumeration of enterococci in water, food and other material.

Preparation Note

Add 76 g to 1 litre distilled water and dissolve by boiling. Dispense into 100 ml portions and sterilize by autoclaving at 121°C for 10 minutes. When ready to use, cool to 50°C and add 1 ml of 1% triphenyltetrazolium chloride solution (Cat. No. 17779) per 100 ml. Do NOT overheat this medium.

Other Notes

Can be prepared and melted by microwave energy[1]; Cultivation and enumeration of streptococci in surface waters[2]

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

B.A. Kenner et al.
Applied Microbiology, 9, 15-15 (1960)
C. Liang et al.
Journal of Food Protection, 51, 577-577 (1988)

Articles

Microbiological assessment of bacteria found in spices, a potent source for food spoilage and pathogens.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service