Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

60459

Sigma-Aldrich

Potassium permanganate

BioUltra, ≥99.0% (RT)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
KMnO4
CAS Number:
Molecular Weight:
158.03
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

product line

BioUltra

assay

≥99.0% (RT)

form

crystals

impurities

insoluble matter, passes filter test
≤0.003% total nitrogen (N)

pH

7-10 (25 °C, 0.1 M in H2O)

solubility

H2O: 0.1 M at 20 °C, clear, violet

anion traces

chloride, chlorate (as Cl-): ≤50 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

As: ≤10 mg/kg
Ca: ≤150 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤10 mg/kg
Cu: ≤5 mg/kg
Fe: ≤50 mg/kg
Mg: ≤100 mg/kg
Na: ≤200 mg/kg
Ni: ≤10 mg/kg
Pb: ≤10 mg/kg
Zn: ≤10 mg/kg

absorption

no transmittance (UV) in H2O at 0.1 M

SMILES string

[K+].[O-][Mn](=O)(=O)=O

InChI

1S/K.Mn.4O/q+1;;;;;-1

InChI key

VZJVWSHVAAUDKD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

application

For volumetric redox analysis, preparation
Potassium permanganate, a strong oxidizing agent, is used to bleach organic compound such as melanin in analytical procedures such as immunostaining protocols and Feulgen-DNA microdensitometry.
Suitable for heterogeneous oxidations.

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Ox. Sol. 2 - Repr. 2 - Skin Corr. 1 - STOT RE 2 Inhalation

target_organs

Brain

Storage Class

5.1B - Oxidizing hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arnaud Beduneau et al.
PloS one, 4(2), e4343-e4343 (2009-02-03)
We posit that the same mononuclear phagocytes (MP) that serve as target cells and vehicles for a host of microbial infections can be used to improve diagnostics and drug delivery. We also theorize that physical and biological processes such as
Darryl B Jones et al.
Water research, 46(17), 5491-5498 (2012-08-15)
In this study, the impacts of three preoxidation strategies [i.e., using potassium permanganate (KMnO(4)), chlorine dioxide (ClO(2)), or hydrogen peroxide (H(2)O(2))] before preformed monochloramine (NH(2)Cl) addition on the formation and speciation of iodinated trihalomethanes (I-THMs) were evaluated at the Br(-)/I(-)
Christopher H Gibbons et al.
Neurology, 72(17), 1479-1486 (2009-04-29)
To evaluate a novel method to quantify the density of nerve fibers innervating sweat glands in healthy control and diabetic subjects, to compare the results to an unbiased stereologic technique, and to identify the relationship to standardized physical examination and
F C Holstege et al.
Methods (San Diego, Calif.), 12(3), 203-211 (1997-07-01)
Open complex formation precedes initiation of transcription by RNA polymerases. In the analysis of transcription initiation from eukaryotic class II promoters, we have used promoter DNA structures that represent intermediates in open complex formation. We describe the preparation and isolation
Jessica M Terry et al.
Talanta, 99, 1051-1056 (2012-09-13)
We have explored the chemiluminescence response of amino acids and related compounds (including the tripeptide glutathione, and disulfides: cystine, homocystine and glutathione disulfide) with several new adaptations of the permanganate and tris(2,2'-bipyridine)ruthenium(III) ([Ru(bipy)(3)](3+)) reagents and a recently developed colloidal manganese(IV)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service