Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

60388

Supelco

Potassium iodate

BioUltra, ≥99.5% (RT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KIO3
CAS Number:
Molecular Weight:
214.00
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

product line

BioUltra

assay

≥99.5% (RT)

form

solid

impurities

insoluble matter, passes filter test
≤0.002% total nitrogen (N)

loss

≤0.05% loss on drying, 110 °C

pH

5.0-8.0 (25 °C, 0.1 M in H2O)

mp

560 °C (lit.)

solubility

H2O: 0.1 M at 20 °C, clear, colorless

density

3.93 g/mL at 25 °C (lit.)

anion traces

bromide, bromate, chloride, chlorate (as Cl-): ≤200 mg/kg
iodide (I-): ≤20 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
Ba: ≤50 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

absorption

cut-off at 276 nm in H2O at 0.1 M

storage temp.

2-8°C

SMILES string

[K+].[O-]I(=O)=O

InChI

1S/HIO3.K/c2-1(3)4;/h(H,2,3,4);/q;+1/p-1

InChI key

JLKDVMWYMMLWTI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Other Notes

Primary standard for iodometry;

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Dryden et al.
Journal of perinatology : official journal of the California Perinatal Association, 27(5), 320-322 (2007-04-25)
Congenital thyrotoxicosis is a rare and potentially fatal illness. We report a case in a preterm infant delivered to a mother known to have autoimmune endocrine disease. Diagnosis was difficult because the infant's presenting symptoms and signs closely resembled congenital
Tatsuya Kawai et al.
Academic radiology, 18(6), 682-689 (2011-03-12)
Evaluation of contrast enhancement of pulmonary lesions with ground-glass attenuation (GGA) is difficult with conventional techniques but might be possible using contrast-mapping images (CMIs) obtained by dual-energy computed tomography. To address this issue, a phantom study was conducted, and this
Mohsen Zeeb et al.
Talanta, 78(2), 584-589 (2009-02-11)
In this work, an on-line system with vapor-phase generation (VPG) and Fourier transform infrared (FTIR) spectrometric detection has been developed as a direct and highly selective analytical technique for the assay of penicillamine (PA). Potassium iodate solution was injected into
Qimeng Li et al.
European journal of nutrition, 51(6), 685-691 (2011-09-23)
The aim of the present feeding trial with iodine was to assess pigs' growth performance and carcass characteristics, the iodine accumulation in tissues, and their influences on the thyroid hormones in plasma. Eighty pigs (33-115 kg body weight) were allotted
Qiang Ren et al.
Journal of trace elements in medicine and biology : organ of the Society for Minerals and Trace Elements (GMS), 22(1), 1-8 (2008-03-06)
Iodine deficiency disorders continue to be a severe problem in many parts of Central Asia, causing delayed mental development and cretinism in indigenous populations. In some areas, iodized salt has not succeeded in controlling this problem. In southern Xinjiang Province

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service