60244
Potassium fluoride on aluminum oxide
extent of labeling: ~5.5 mmol/g F- loading
About This Item
Recommended Products
extent of labeling
~5.5 mmol/g F- loading
SMILES string
[F-].[K+]
InChI
1S/FH.K/h1H;/q;+1/p-1
InChI key
NROKBHXJSPEDAR-UHFFFAOYSA-M
Storage Class
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service