60031
Potassium ionophore I - cocktail A
Selectophore™
About This Item
Recommended Products
product line
Selectophore™
composition
1,2-dimethyl-3-nitrobenzene, 25.0 wt. %
potassium ionophore I, 5.0 wt. %
potassium tetrakis(4-chlorophenyl)borate, 2.0 wt. %
sebacic acid dibutylester, 68.0 wt. %
General description
Packaging
Other Notes
Legal Information
replaced by
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 2 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 2
Storage Class
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service