Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

59919

Supelco

Betulin 3,28-diacetate

analytical standard

Synonym(s):

Betulin diacetate, Betulinol diacetate, Lup-20(30)-ene-3β,28-diol diacetate, NSC 38876

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H54O4
CAS Number:
Molecular Weight:
526.79
Beilstein/REAXYS Number:
2067908
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤2.0% water

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

[H][C@]12[C@@]([C@@](CC[C@]3(COC(C)=O)[C@]4([H])[C@H](C(C)=C)CC3)(C)[C@]4([H])CC2)(C)CC[C@@]5([H])C(C)(C)[C@@H](OC(C)=O)CC[C@@]51C

InChI

1S/C34H54O4/c1-21(2)24-12-17-34(20-37-22(3)35)19-18-32(8)25(29(24)34)10-11-27-31(7)15-14-28(38-23(4)36)30(5,6)26(31)13-16-33(27,32)9/h24-29H,1,10-20H2,2-9H3/t24-,25+,26-,27+,28-,29+,31-,32+,33+,34+/m0/s1

InChI key

MIROITGPMGDCGI-MQXQNARFSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service