Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

58833

Sigma-Aldrich

Diethyl isocyanomethylphosphonate

purum, ≥97.0% (GC)

Synonym(s):

Isocyanomethylphosphonic acid diethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CNCH2P(O)(OC2H5)2
CAS Number:
Molecular Weight:
177.14
Beilstein/REAXYS Number:
4674920
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

grade

purum

assay

≥97.0% (GC)

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.433 (lit.)

bp

84-87 °C/0.1 mmHg (lit.)

density

1.105 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCOP(=O)(C[N+]#[C-])OCC

InChI

1S/C6H12NO3P/c1-4-9-11(8,6-7-3)10-5-2/h4-6H2,1-2H3

InChI key

ICURVXBGGDVHDT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Copper-catalyzed cycloaddition reactions
  • Preparation of orally bioavailable diamide prodrugs that lower glucose in diabetic animals through inhibition of fructose-1,6-bisphosphatase
  • Synthesis of sterol analogs as antifungal agents against plant pathogens
  • Asymmetric synthesis of steroidal 2,5-diketopiperazines prepared via isocyanide chemistry based on stereoselective Ugi four component condensation and subsequent cyclocondensation reactions
  • The Passerini reaction

Other Notes

Prep. of phosphonic acid analogs of amino acids[1][2][3]; Synthesis of aldehydes from carbonyl compounds[4][5]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. Stoelwinder et al
The Journal of Organic Chemistry, 57, 2249-2249 (1992)
U. Schollkopf et al.
Synthesis, 1033-1033 (1984)
B. Kohler et al.
Liebigs Ann. Chem., 267-267 (1987)
J. Moskal et al.
Rec. Trav. Chim., 106, 137-137 (1987)
J. Rachon
Synthesis, 219-219 (1984)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service