58700
Ethyl isobutyrylacetate
purum, ≥95.0% (GC)
Synonym(s):
Ethyl 4-methyl-3-oxopentanoate
About This Item
grade
purum
assay
≥95.0% (GC)
refractive index
n20/D 1.425 (lit.)
n20/D 1.426
bp
173 °C (lit.)
mp
−9 °C (lit.)
density
0.98 g/mL at 25 °C (lit.)
SMILES string
CCOC(=O)CC(=O)C(C)C
InChI
1S/C8H14O3/c1-4-11-8(10)5-7(9)6(2)3/h6H,4-5H2,1-3H3
InChI key
XCLDSQRVMMXWMS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
signalword
Warning
hcodes
Hazard Classifications
Flam. Liq. 3
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
111.2 °F - closed cup
flash_point_c
44 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service