Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

58390

Sigma-Aldrich

Isobutyric anhydride

purum, ≥98.0% (GC)

Synonym(s):

2-Methylpropionic anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCO2COCH(CH3)2
CAS Number:
Molecular Weight:
158.19
Beilstein/REAXYS Number:
386267
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

5.45 (vs air)

vapor pressure

10 mmHg ( 67 °C)

grade

purum

assay

≥98.0% (GC)

autoignition temp.

624 °F

expl. lim.

1.09 %, 87 °F
7.7 %, 127 °F

refractive index

n20/D 1.406 (lit.)
n20/D 1.406

bp

182 °C (lit.)

mp

−56 °C (lit.)

density

0.954 g/mL at 25 °C (lit.)

SMILES string

CC(C)C(=O)OC(=O)C(C)C

InChI

1S/C8H14O3/c1-5(2)7(9)11-8(10)6(3)4/h5-6H,1-4H3

InChI key

LSACYLWPPQLVSM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

This product has been replaced by 245771-ALDRICH | Isobutyric anhydride 97%

replaced by

Product No.
Description
Pricing

pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Skin Corr. 1B

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

154.4 °F - closed cup

flash_point_c

68 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service