Key Documents
58166
Supelco
Ion-Selective Optode membranes
About This Item
UNSPSC Code:
26111700
Quality Level
General description
English translation and revised edition of the German monograph (Cat. No. 58165) describing theory, preparation and application of ion-selective optode membranes as well as recent developments in this field.With 237 references.
Visit our Sensor Applications portal to learn more.
Other Notes
K. Seiler, 1993, 114 pp.; edited by Sigma-Aldrich Chemie GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)