Well developed granules of volutin (polyphosphate) may be seen in unstained wet preparations as round refractive bodies within the bacterial cytoplasm. With basic dye, they tend to stain more strongly than the rest of the bacterium, and with methylene blue solution (Cat. No. 01363) they stain metachromatically, a reddish purple color.
Application
Neutral Red, a histological vital stain, is used as a stain and counterstain. It is used to stain Golgi apparatus in cells and Nissi granules in neurons. As a pH sensitive dye, neutral red stains lysosomes red within cells.
Components
Composition: Neutral red 1.0 g/l, Acetic acid 2 ml/l in water
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
Although MTT is widely used to assess cytotoxicity and cell viability, the precise localization of its reduced formazan product is still unclear. In the present study the localization of MTT formazan was studied by direct microscopic observation of living HeLa
Chemical research in toxicology, 25(8), 1662-1674 (2012-06-27)
Enniatins are cyclic hexadepsipeptidic mycotoxins with ionophoric, antibiotic, and insecticidal activity. Enniatin B (EnnB), the most important analogue, is produced by many Fusarium species and is a common contaminant in grain-based foods. The compound's cytotoxic potential has been shown in
Microscopy research and technique, 76(2), 126-132 (2012-10-31)
The aim of this randomized crossover investigation was to assess the biofilm formation on two titanium and one zirconia substrates in relation to the topography and surface roughness of the materials. Twenty-four discs specimens for each evaluated material (Machined pure
Toxicology in vitro : an international journal published in association with BIBRA, 27(2), 760-767 (2012-12-25)
When in vitro test systems are evaluated for assessment of the toxicity of chemical compounds, particular efforts are made to mimic the in vivo reality as close as possible. Cellular models with appropriate metabolic competence, i.e. with the potency to
This study introduces a novel and simple method to covalently graft neutral red (NR) onto carbon surfaces based on spontaneous reduction of in situ generated NR diazonium salts. Immobilization of neutral red on carbon surface was achieved by immersing carbon
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.