Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

57680

Sigma-Aldrich

2-Iodoaniline

purum, ≥98.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
IC6H4NH2
CAS Number:
Molecular Weight:
219.02
Beilstein/REAXYS Number:
2204899
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

assay

≥98.0% (GC)

form

solid

mp

55-57 °C
55-58 °C (lit.)

SMILES string

Nc1ccccc1I

InChI

1S/C6H6IN/c7-5-3-1-2-4-6(5)8/h1-4H,8H2

InChI key

UBPDKIDWEADHPP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service