Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

57660

Sigma-Aldrich

Iodine

purum p.a., ≥99.5% (RT)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
I2
CAS Number:
Molecular Weight:
253.81
Beilstein/REAXYS Number:
3587194
EC Number:
MDL number:
UNSPSC Code:
12141916
PubChem Substance ID:

vapor density

9 (vs air)

vapor pressure

0.31 mmHg ( 25 °C)
1 mmHg ( 38.7 °C)

grade

purum p.a.

assay

≥99.5% (RT)

resistivity

1.3E15 μΩ-cm

bp

184 °C (lit.)

mp

113 °C (lit.)

anion traces

chlorine and bromine (as Cl-): ≤200 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤100 mg/kg
Na: ≤100 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

II

InChI

1S/I2/c1-2

InChI key

PNDPGZBMCMUPRI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

target_organs

Respiratory system, Thyroid

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A simple spectrophotometric determination of total β-carotene in food additives with varying E/Z-isomer ratios using an isobestic wavelength.
Schierle J, et al.
European Food Research and Technology, 215(3), 268-274 (2002)
Eagleson M.
Concise Encyclopedia Chemistry, 543-543 (1994)
A spectrophotometric investigation of the interaction of iodine with aromatic hydrocarbons.
Benesi HA and Hildebrand JH.
Journal of the American Chemical Society, 71(8), 2703-2707 (1949)
Michael B Zimmermann
Thyroid : official journal of the American Thyroid Association, 17(9), 829-835 (2007-10-25)
Iodine is required for the production of thyroid hormones, which are essential for normal brain development, and the fetus, newborn, and young child are particularly vulnerable to iodine deficiency. The iodine requirement increases during pregnancy and recommended intakes are in
Sarah C Bath et al.
Lancet (London, England), 382(9889), 331-337 (2013-05-28)
As a component of thyroid hormones, iodine is essential for fetal brain development. Although the UK has long been considered iodine replete, increasing evidence suggests that it might now be mildly iodine deficient. We assessed whether mild iodine deficiency during

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service