Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

56907

Supelco

Rubidium Ionophore I

Selectophore, function tested

Synonym(s):

2,5,8,10,13,16-Hexaoxa-1(1,8)-anthraquinona-9(2,2′)-1,1′-binaphthalinacyclohexadecaphane, Anthrachinono[1,9a,9,8a,8-stuv]-dinaphtho[2,1-h:1′,2′-j]-1,4,7,12,15,18-hexaoxacyclotricosene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H34O8
CAS Number:
Molecular Weight:
666.71
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

product line

Selectophore

form

solid

SMILES string

O=C1c2cccc3OCCOCCOc4ccc5ccccc5c4-c6c(OCCOCCOc7cccc1c7C(=O)c23)ccc8ccccc68

InChI

1S/C42H34O8/c43-41-31-11-5-13-33-39(31)42(44)40-32(41)12-6-14-34(40)48-24-20-46-22-26-50-36-18-16-28-8-2-4-10-30(28)38(36)37-29-9-3-1-7-27(29)15-17-35(37)49-25-21-45-19-23-47-33/h1-18H,19-26H2

InChI key

BTGQLABSOYZCLX-UHFFFAOYSA-N

General description

Visit our Sensor Applications portal to learn more.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service