56880
1-Indanone
purum, ≥98.0% (GC)
Synonym(s):
α-Hydrindone
About This Item
Recommended Products
grade
purum
assay
≥98.0% (GC)
bp
243-245 °C (lit.)
mp
38-40 °C (lit.)
39-41 °C
density
1.103 g/mL at 25 °C (lit.)
SMILES string
O=C1CCc2ccccc12
InChI
1S/C9H8O/c10-9-6-5-7-3-1-2-4-8(7)9/h1-4H,5-6H2
InChI key
QNXSIUBBGPHDDE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
233.0 °F - closed cup
flash_point_c
111.67 °C - closed cup
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service