Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

56869

Supelco

Triisopropyl phosphate

analytical standard

Synonym(s):

NSC 46370, NSC 62275, Phosphoric acid triisopropyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PO(OCH(CH3)2)3
CAS Number:
Molecular Weight:
224.23
EC Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥95.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.404-1.410
n20/D 1.4070 (lit.)

bp

224 °C (lit.)

density

0.970 g/mL at 25 °C (lit.)

application(s)

environmental

SMILES string

CC(C)OP(=O)(OC(C)C)OC(C)C

InChI

1S/C9H21O4P/c1-7(2)11-14(10,12-8(3)4)13-9(5)6/h7-9H,1-6H3

InChI key

OXFUXNFMHFCELM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triisopropyl phosphate is an alkyl phosphate commonly found in petroleum and crude oil products.[1]

Application

Triisopropyl phosphate may be used as an analytical reference standard for the quantification of the analyte in petroleum samples[2][3] and crude oil[1] using two-dimensional gas chromatography technique.

signalword

Warning

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

73.4 °F - closed cup

flash_point_c

23 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Profiling Alkyl Phosphates in Industrial Petroleum Samples by Comprehensive Two-Dimensional Gas Chromatography with Nitrogen Phosphorus Detection (GC? GC?NPD), Post-column Deans Switching, and Concurrent Backflushing
Nizio DK and Harynuk JJ
Energy and Fuels, 28(3), 1709-1716 (2014)
Analysis of alkyl phosphates in petroleum samples by comprehensive two-dimensional gas chromatography with nitrogen phosphorus detection and post-column Deans switching
Nizio DK and Harynuk JJ
Journal of Chromatography A, 1252, 171-176 (2012)
The analysis of alkyl phosphates in nitrogen-rich crude oils using GC? GC-NPD with a polar/apolar column configuration
Stevenson MJAK and Harynuk JJ
Analytical Methods : Advancing Methods and Applications, 9(36), 5301-5309 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service