55905
Iodine Solution (0.05M in THF/H2O/pyridine 70:20:10)
suitable for DNA synthesis
Synonym(s):
Iodine solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
I2
CAS Number:
Molecular Weight:
253.81
Beilstein/REAXYS Number:
3587194
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
technique(s)
DNA synthesis: suitable
density
0.95 g/mL at 20 °C
SMILES string
II
InChI
1S/I2/c1-2
InChI key
PNDPGZBMCMUPRI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Oxidizing solution in the phosphote amide triester method of oligonucleotide synthesis
Application
suitable for the use on Milligen 7500 Synthesizers
Preparation Note
filtered with a 0.1 μm filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Yanfeng Zhang et al.
Experimental and therapeutic medicine, 9(4), 1351-1356 (2015-03-18)
Oxytocin (OT) neurons localized in the paraventricular nucleus (PVN) and supraoptic nucleus (SON) send fibers to the brain and spinal cord. While most previous studies have looked at the role of OT in chronic pain, few have investigated the role
Hélène Guet-Revillet et al.
Emerging infectious diseases, 20(12), 1990-1998 (2014-11-25)
Hidradenitis suppurativa (HS) is a skin disease characterized by recurrent nodules or abscesses and chronic suppurating lesions. In the absence of clear pathophysiology, HS is considered to be an inflammatory disease and has no satisfactory medical treatment. Recently, prolonged antimicrobial
Hakimeh Zali et al.
Iranian journal of pharmaceutical research : IJPR, 14(1), 291-302 (2015-01-07)
Different treatment strategies of Alzheimer's disease (AD) are being studied for treating or slowing the progression of AD. Many pharmaceutically important regulation systems operate through proteins as drug targets. Here, we investigate the drug target proteins in beta-amyloid (Aβ) injected
Ming-Li Chou et al.
PloS one, 9(6), e99145-e99145 (2014-06-20)
Clinical-grade ex vivo expansion of corneal endothelial cells can increase the availability of corneal tissues for transplantation and treatment of corneal blindness. However, these cells have very limited proliferative capacity. Successful propagation has required so far to use very complex
Paul J Austin et al.
PloS one, 10(4), e0124755-e0124755 (2015-04-24)
Allodynia, hyperalgesia and spontaneous pain are cardinal sensory signs of neuropathic pain. Clinically, many neuropathic pain patients experience affective-motivational state changes, including reduced familial and social interactions, decreased motivation, anhedonia and depression which are severely debilitating. In earlier studies we
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service