Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

55862

Supelco

4-(4-Hydroxy-4-methylpentyl)-3-cyclohexene-1-carboxaldehyde

mixture of isomers, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H22O2
CAS Number:
Molecular Weight:
210.31
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥95.0% (sum of isomers, GC)
65.0-85.0% (GC)

shelf life

limited shelf life, expiry date on the label

concentration

15.0-35.0% (minor component, GC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

density

0.995 g/mL at 20 °C

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

[H]C(=O)C1CCC(CCCC(C)(C)O)=CC1

InChI

1S/C13H22O2/c1-13(2,15)9-3-4-11-5-7-12(10-14)8-6-11/h5,10,12,15H,3-4,6-9H2,1-2H3

InChI key

ORMHZBNNECIKOH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

275.2 °F

flash_point_c

135.1 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service