Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

55836

Supelco

Quercetin 3-O-rutinoside-7-O-glucoside

analytical standard

Synonym(s):

3,3′,4′,5,7-Pentahydroxyflavone 3-rutinoside 7-glucoside, 3-[[6-O-(6-Deoxy-α-L-mannopyranosyl)-β-D-glucopyranosyl]oxy]-2-(3,4-dihydroxyphenyl)-7-(β-D-glucopyranosyloxy)-5-hydroxy-4H-1-benzopyran-4-one, Quercetin 3-rhamnoglucoside 7-glucoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H40O21
CAS Number:
Molecular Weight:
772.66
UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

assay

≥90.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

food and beverages

format

neat

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Quercetin 3-O-rutinoside-7-O-glucoside is a flavonoid compound found in mulberry leaves and fruits.[1]

Application

Quercetin 3-O-rutinoside-7-O-glucoside may be used as a reference standard for the determination of the analyte in Withania somnifera aerial parts by high performance liquid chromatography method.[2]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and validation of high performance liquid chromatography method for simultaneous estimation of flavonoid glycosides in Withania somnifera aerial parts.
Mundkinajeddu D, et al.
ISRN Analytical Chemistry, 2014(5), 1789-1796 (2014)
Qualitative and quantitative analysis of flavonoids from 12 species of Korean mulberry leaves.
Ju WT, et al.
Journal of Food Science and Technology, 55(5), 1789-1796 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service