Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

55645

Supelco

D-2-Aminoadipic acid

analytical standard

Synonym(s):

(R)-2-Aminohexanedioic acid, D-Homoglutamic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2C(CH2)3CH(NH2)CO2H
CAS Number:
Molecular Weight:
161.16
Beilstein/REAXYS Number:
1724347
MDL number:
UNSPSC Code:
41116107
Product 55645 is not currently sold in your country. Contact Technical Service

grade

analytical standard

assay

≥98% (TLC)

form

solid

optical activity

[α]25/D -24.5±1.5°, c = 2 in 5 M HCl

shelf life

limited shelf life, expiry date on the label

mp

208-210 °C (lit.)

solubility

1 M HCl: 50 mg/mL, clear, colorless

SMILES string

N[C@H](CCCC(O)=O)C(O)=O

InChI

1S/C6H11NO4/c7-4(6(10)11)2-1-3-5(8)9/h4H,1-3,7H2,(H,8,9)(H,10,11)/t4-/m1/s1

InChI key

OYIFNHCXNCRBQI-SCSAIBSYSA-N

Looking for similar products? Visit Product Comparison Guide

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service