Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

55571

Supelco

Assay of arsenic, simile substance

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113319

application(s)

food and beverages
general analytical

Quality Level

Analysis Note

10 mg/L arsenic in 0.5 N potassium hydroxide solution

pictograms

Health hazardCorrosion

signalword

Danger

Hazard Classifications

Carc. 1A - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service