Skip to Content
MilliporeSigma

55500

DL-5-Hydroxylysine hydrochloride

purum, mixture of DL- and DL-allo-form, ≥97.0% (AT)

Synonym(s):

2,6-Diamino-5-hydroxycaproic acid hydrochloride, 2,6-Diamino-5-hydroxyhexanoic acid hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
NH2CH2CH(OH)CH2CH2CH(NH2)COOH · HCl
CAS Number:
Molecular Weight:
198.65
EC Number:
236-168-9
UNSPSC Code:
12352200
PubChem Substance ID:
Beilstein/REAXYS Number:
3914368
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

purum

assay

≥97.0% (AT)

mp

225 °C (dec.) (lit.), 230 °C (dec.)

SMILES string

Cl.NCC(O)CCC(N)C(O)=O

InChI

1S/C6H14N2O3.ClH/c7-3-4(9)1-2-5(8)6(10)11;/h4-5,9H,1-3,7-8H2,(H,10,11);1H

InChI key

MJXVOTKVFFAZQJ-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



A Rahman et al.
Plant & cell physiology, 41(1), 1-9 (2000-04-06)
Chromosaponin I (CSI), a triterpenoid saponin isolated from pea, stimulates the growth of roots in Arabidopsis thaliana seedlings on wetted filter paper in the light for 14 d. The growth rates of roots in Columbia (Col) and Landsberg erecta (Ler)
Nenggang Zhang et al.
Physiologia plantarum, 116(3), 383-388 (2003-01-25)
We studied the mode of action of 4,4,4-trifluoro-3- (indole-3-) butyric acid (TFIBA), a recently described root growth stimulator, on primary root growth of Lactuca sativa L. seedlings. TFIBA (100 micromoles) promoted elongation of primary roots by 40% in 72 h
J-S Han et al.
Plant cell reports, 23(5), 291-296 (2004-09-15)
Using cotyledon explants excised from seedlings germinated in vitro, an efficient plant regeneration system via organogenesis was established for bottle gourd (Lagenaria siceraria Standl.). Maximum shoot regeneration was obtained when the proximal parts of cotyledons from 4-day-old seedlings were cultured