Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

55381

Supelco

2-Methyl-2-thiazoline

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7NS
CAS Number:
Molecular Weight:
101.17
Beilstein/REAXYS Number:
104274
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

grade

analytical standard

assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

impurities

≤5.0% water

refractive index

n20/D 1.518-1.522
n20/D 1.520 (lit.)

bp

144.0-145.0 °C (lit.)

mp

-101.0 °C (lit.)

density

1.067 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

SMILES string

CC1=NCCS1

InChI

1S/C4H7NS/c1-4-5-2-3-6-4/h2-3H2,1H3

InChI key

JUIQOABNSLTJSW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Flame

signalword

Warning

hcodes

pcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

98.6 °F - closed cup

flash_point_c

37 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weili Tian et al.
Communications biology, 3(1), 49-49 (2020-02-01)
Protecting the nervous system from chronic effects of physical and chemical stress is a pressing clinical challenge. The obligate pro-degenerative protein Sarm1 is essential for Wallerian axon degeneration. Thus, blocking Sarm1 function is emerging as a promising neuroprotective strategy with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service