Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

55329

Supelco

L-Alanine hydrochloride solution

100 mM amino acid in 0.1 M HCl, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H7NO2 · HCl
CAS Number:
Molecular Weight:
125.55
Beilstein/REAXYS Number:
4296703
MDL number:
UNSPSC Code:
85151701
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

form

liquid

analyte chemical class(es)

amino acids, peptides, proteins

concentration

100 mM amino acid in 0.1 M HCl

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

colorless

application(s)

food and beverages

format

single component solution

storage temp.

2-8°C

SMILES string

Cl.C[C@H](N)C(O)=O

InChI

1S/C3H7NO2.ClH/c1-2(4)3(5)6;/h2H,4H2,1H3,(H,5,6);1H/t2-;/m0./s1

InChI key

ILYVXUGGBVATGA-DKWTVANSSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Elliott S Kerrin et al.
Analytical and bioanalytical chemistry, 409(6), 1481-1491 (2016-12-03)
Recent reports of the widespread occurrence of the neurotoxin β-N-methylamino-L-alanine (BMAA) in cyanobacteria and particularly seafood have raised concerns for public health. LC-MS/MS is currently the analytical method of choice for BMAA determinations but incomplete separation of isomeric and isobaric

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service