Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

55200

Sigma-Aldrich

D-(−)-Pantolactone

puriss., ≥99.0% (T)

Synonym(s):

(R)-(−)-α-Hydroxy-β,β-dimethyl-γ-butyrolactone, (R)-(−)-β,β-Dimethyl-α-hydroxy-γ-butyrolactone, (R)-(−)-Pantolactone, Pantoic acid γ-lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O3
CAS Number:
Molecular Weight:
130.14
Beilstein/REAXYS Number:
80957
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

grade

puriss.

assay

≥99.0% (T)

form

solid

optical activity

[α]20/D −51±2°, c = 3% in H2O

bp

120-122 °C/15 mmHg (lit.)

mp

89-92 °C
91 °C (lit.)

storage temp.

2-8°C

SMILES string

CC1(C)COC(=O)[C@@H]1O

InChI

1S/C6H10O3/c1-6(2)3-9-5(8)4(6)7/h4,7H,3H2,1-2H3/t4-/m0/s1

InChI key

SERHXTVXHNVDKA-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Use as chiral building block: reduction to the triol; Amidation; Convenient acces to two enantiomeric oxiranes: S(+)- and R(-)-2,2-dimethyl-3,4-epoxybutanol

replaced by

Product No.
Description
Pricing

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G.C. Fischer et al.
The Journal of Organic Chemistry, 50, 2011-2011 (1985)
R.E. Dolle et al.
Journal of the American Chemical Society, 107, 1691-1691 (1985)
P. Lavallee et al.
Tetrahedron Letters, 27, 679-679 (1986)
Dayong Si et al.
Applied microbiology and biotechnology, 95(2), 431-440 (2012-03-09)
The 1,2-propanediol (1,2-PD) inducible membrane-bound L-pantoyl lactone (L-PL) dehydrogenase (LPLDH) has been isolated from Rhodococcus erythropolis AKU2103 (Kataoka et al. in Eur J Biochem 204:799, 1992). Based on the N-terminal amino acid sequence of LPLDH and the highly conserved amino
Zhi-Hui Peng et al.
Organic letters, 4(17), 2945-2948 (2002-08-17)
[reaction: see text] The [3 + 2] annulation reaction of allylsilane 1 with an alpha-keto ester provided the highly substituted tetrahydrofuran 2 as a single diastereomer in high yield. The synthesis of (+/-)-5-epi-citreoviral and (+/-)-citreoviral has been accomplished with this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service