Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

55187

Supelco

2,5-Dimethyl-4-methoxy-3(2H)-furanone

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10O3
CAS Number:
Molecular Weight:
142.15
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.476-1.480
n20/D 1.478 (lit.)

density

1.097 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

SMILES string

COC1=C(C)OC(C)C1=O

InChI

1S/C7H10O3/c1-4-6(8)7(9-3)5(2)10-4/h4H,1-3H3

InChI key

SIMKGHMLPVDSJE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

167.0 °F - closed cup

flash_point_c

75 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service