Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

55135

Sigma-Aldrich

Cyclohexane - ethyl acetate 1:1 (v/v) mixture

suitable for pesticide residue analysis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12190000
NACRES:
NA.06
technique(s):
PET imaging: suitable

product line

PESTANAL®

Quality Level

form

liquid

composition

cyclohexane, 49.5-50.5% GC
ethyl acetate, 49.5-50.5% GC

technique(s)

PET imaging: suitable

impurities

≤0.01% water
≤5 ng/L lindane

evapn. residue

≤0.001%

suitability

suitable for pesticide residue analysis

Application

Suitable for sample cleanup and/or extraction when performing pesticide residue analysis[1][2].

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

-0.4 °F

flash_point_c

-18 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

DFG Deutsche Forschungsgemeinschaft
Manual of Pesticide Residue Analysis, I (1987)
DFG Deutsche Forschungsgemeinschaft
Manual of Pesticide Residue Analysis, II (1992)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service