55070
8-Quinolinol
puriss. p.a., ACS reagent, for the detection and determination of Al, Mg and others, ≥99.0% (NT)
Synonym(s):
8-Hydroxyquinoline, 8-Oxychinolin, Oxine
About This Item
Recommended Products
grade
ACS reagent
puriss. p.a.
assay
≥99.0% (NT)
quality
for the detection and determination of Al, Mg and others
mp
72.5-74 °C
72.5-74.0 °C
antibiotic activity spectrum
fungi
mode of action
DNA synthesis | interferes
enzyme | inhibits
SMILES string
Oc1cccc2cccnc12
InChI
1S/C9H7NO/c11-8-5-1-3-7-4-2-6-10-9(7)8/h1-6,11H
InChI key
MCJGNVYPOGVAJF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
signalword
Danger
Hazard Classifications
Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 1B - Skin Sens. 1
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Faceshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service