Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

54957

Sigma-Aldrich

Methyl (R)-3-hydroxybutyrate

puriss., ≥99.0% (sum of enantiomers, GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH2CO2CH3
CAS Number:
Molecular Weight:
118.13
Beilstein/REAXYS Number:
6130687
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

grade

puriss.

assay

≥99.0% (sum of enantiomers, GC)

optical activity

[α]20/D −50±2°, c = 1.3% in chloroform stab. with amylenes

optical purity

enantiomeric ratio: ≥98:2 (GC)

refractive index

n20/D 1.421 (lit.)
n20/D 1.422

bp

56-58 °C/11 mmHg (lit.)

density

1.055 g/mL at 20 °C (lit.)

SMILES string

COC(=O)C[C@@H](C)O

InChI

1S/C5H10O3/c1-4(6)3-5(7)8-2/h4,6H,3H2,1-2H3/t4-/m1/s1

InChI key

LDLDJEAVRNAEBW-SCSAIBSYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Important optically active compound for the preparation of various bifunctional building blocks [1][2][3][4]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

163.4 °F - closed cup

flash_point_c

73 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

First total synthesis of grahamimycin A.
Y Kobayashi et al.
The Journal of organic chemistry, 65(21), 7221-7224 (2000-10-14)
R M Garbaccio et al.
Organic letters, 2(20), 3127-3129 (2000-09-29)
A concise, stereospecific synthesis of radicicol dimethyl ether is presented. The strategy relies on a convergent three-stage assembly of the 14-membered lactone which has, as a key transformation, a novel ring-forming metathesis reaction utilizing a vinyl epoxide.
A new synthesis of a key intermediate of beta-lactam antibiotics via diastereoselective alkylation of beta-hydroxy ester.
W H Ham et al.
The Journal of organic chemistry, 65(24), 8372-8374 (2000-12-02)
A. Fischli et al.
Modern Synthetic Methods, 2, 269-269 (1980)
Xiao-Hong Chen et al.
PloS one, 9(4), e94543-e94543 (2014-04-18)
A novel carbonyl reductase (AcCR) catalyzing the asymmetric reduction of ketones to enantiopure alcohols with anti-Prelog stereoselectivity was found in Acetobacter sp. CCTCC M209061 and enriched 27.5-fold with an overall yield of 0.4% by purification. The enzyme showed a homotetrameric

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service