Skip to Content
MilliporeSigma

54860

4-Hydroxybenzyl alcohol

purum, ≥98.5% (HPLC)

Synonym(s):

NSC 227926

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
HOC6H4CH2OH
CAS Number:
Molecular Weight:
124.14
EC Number:
210-768-0
UNSPSC Code:
12352200
PubChem Substance ID:
Beilstein/REAXYS Number:
1858967
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

purum

assay

≥98.5% (HPLC)

reaction suitability

reagent type: cross-linking reagent

mp

114-122 °C

SMILES string

OCc1ccc(O)cc1

InChI

1S/C7H8O2/c8-5-6-1-3-7(9)4-2-6/h1-4,8-9H,5H2

InChI key

BVJSUAQZOZWCKN-UHFFFAOYSA-N



Still not finding the right product?

Explore all of our products under 4-Hydroxybenzyl alcohol


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Byung-Wook Kim et al.
Journal of ethnopharmacology, 139(2), 549-557 (2011-12-14)
Gastrodia elata Blume (Orchidaceae, GE) a traditional plant in Oriental countries is known for its enormous benefits to treat headaches, dizziness, vertigo and convulsive illnesses. In the present study, the ethnopharmacological role of GE in neuroinflammation mediated by activated microglia
Jia-Shun Gong et al.
Yao xue xue bao = Acta pharmaceutica Sinica, 41(10), 963-966 (2006-12-23)
To investigate the production of p-hydroxymethylphenol-beta-D-glucoside (gastrodin) through biotransformation by plant cell suspension cultures. Using cell suspension cultures of Datura stramonium to convert the exogenous p-hydroxybenzaldehyde into gastrodin was conducted and the converted compounds were separated with a combination of
Niloufar Choubdar et al.
Journal of food science, 75(4), C341-C345 (2010-06-16)
Escherichia coli O157:H7 can survive the dry fermented sausage manufacturing process. Compounds generated by enzymatic hydrolysis of glucosinolates present in ground mustard have shown potential antibacterial properties against E. coli O157:H7. In the present study, supercritical fluid chromatography (SFC) was



Global Trade Item Number

SKUGTIN
338923-10G04061826745649
338923-50G04061837581106