Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
HOC6H4CH2OH
CAS Number:
Molecular Weight:
124.14
EC Number:
210-633-6
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
2041500
MDL number:
grade
purum
assay
≥98.0% (GC)
mp
69-72 °C
SMILES string
OCc1cccc(O)c1
InChI
1S/C7H8O2/c8-5-6-2-1-3-7(9)4-6/h1-4,8-9H,5H2
InChI key
OKVJCVWFVRATSG-UHFFFAOYSA-N
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.