About This Item
Recommended Products
grade
puriss.
assay
≥99.0% (T)
form
solid
mp
213-217 °C (lit.)
214-216 °C
solubility
methanol: 0.5 g/10 mL, clear, colorless
SMILES string
OC(=O)c1ccc(O)cc1
InChI
1S/C7H6O3/c8-6-3-1-5(2-4-6)7(9)10/h1-4,8H,(H,9,10)
InChI key
FJKROLUGYXJWQN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1 - STOT SE 3
target_organs
Respiratory system
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service