Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

54461

Sigma-Aldrich

HEPES

≥99.0% (titration), powder or crystals

Synonym(s):

4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H18N2O4S
CAS Number:
Molecular Weight:
238.30
Beilstein/REAXYS Number:
883043
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:

Product Name

HEPES, ≥99.0% (T)

assay

≥99.0% (T)

form

powder or crystals

ign. residue

≤0.1% (as SO4)

loss

≤0.5% loss on drying, 110 °C

pH

5.0-6.5 (25 °C, 238 g/L)

useful pH range

6.8-8.2

pKa (25 °C)

7.5

solubility

water: 2.38 g/10 mL, clear to slightly hazy, colorless to almost colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤300 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤100 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

OCCN1CCN(CC1)CCS(O)(=O)=O

InChI

1S/C8H18N2O4S/c11-7-5-9-1-3-10(4-2-9)6-8-15(12,13)14/h11H,1-8H2,(H,12,13,14)

InChI key

JKMHFZQWWAIEOD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Biological buffer[1][2]

replaced by

Product No.
Description
Pricing

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P M Brannon et al.
In vitro cellular & developmental biology : journal of the Tissue Culture Association, 21(1), 6-14 (1985-01-01)
Rat pancreatic acinar cells were isolated and cultured in Ham's F12 medium with 15% bovine calf serum. Caerulein, insulin, somatostatin, and dexamethasone (DEX) had no effect on intracellular or secreted amylase in these cultured cells. A serum-free medium, using Waymouth's
Sietse T Braakman et al.
Experimental eye research, 127, 224-235 (2014-08-17)
The bulk of aqueous humor passing through the conventional outflow pathway must cross the inner wall endothelium of Schlemm's canal (SC), likely through micron-sized transendothelial pores. SC pore density is reduced in glaucoma, possibly contributing to obstructed aqueous humor outflow
P Dey et al.
Oncogene, 33(33), 4213-4225 (2013-10-01)
Estrogen receptor β (ERβ) is emerging as a critical factor in understanding prostate cancer biology. Although reduced in prostate cancer above Gleason grade 3, ERβ is a potential drug target at the initial stage of the disease. In human prostate
Frédéric Perros et al.
Journal of the American College of Cardiology, 65(7), 668-680 (2015-02-14)
Endothelial cell (EC) dysfunction plays a central role in the pathogenesis of pulmonary arterial hypertension (PAH), promoting vasoconstriction, smooth muscle proliferation, and inflammation. This study sought to test the hypothesis that nebivolol, a β1-antagonist and β2,3-agonist, may improve PAH and
Alexandre Albanese et al.
ACS nano, 8(6), 5515-5526 (2014-05-07)
A nanoparticle's physical and chemical properties at the time of cell contact will determine the ensuing cellular response. Aggregation and the formation of a protein corona in the extracellular environment will alter nanoparticle size, shape, and surface properties, giving it

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service