Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

54110

Supelco

Hydrogen sulfite ionophore I

Selectophore

Synonym(s):

ETH 5444, Octadecyl 4-formylbenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H42O3
CAS Number:
Molecular Weight:
402.61
Beilstein/REAXYS Number:
6637746
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

product line

Selectophore

Quality Level

SMILES string

[H]C(=O)c1ccc(cc1)C(=O)OCCCCCCCCCCCCCCCCCC

InChI

1S/C26H42O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-22-29-26(28)25-20-18-24(23-27)19-21-25/h18-21,23H,2-17,22H2,1H3

InChI key

DSTQLLFFZIANJD-UHFFFAOYSA-N

General description

Visit our Sensor Applications portal to learn more.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Ionophore used in Chromoionophore III (ETH 5350, Cat. No. 27088) based polymeric membrane sensor for hydrogen sulfite[1]

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M. Kuratli et al.
Analytical Chemistry, 66, 85-85 (1994)
Mariana Raposo et al.
Sensors (Basel, Switzerland), 20(2) (2020-01-23)
Out of control proliferation of toxic phytoplankton, called harmful algal blooms (HABs), have a significant economic impact on bivalve aquaculture and harvesting in coastal waters. Some phytotoxins, such as paralytic shellfish toxins (PSTs), are of concern due to the life-threatening

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service