Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

53490

Sigma-Aldrich

1-Methylpiperidine solution

BioReagent, suitable for protein sequencing, ~40% in 1-butanol

Synonym(s):

N-Methylpiperidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13N
CAS Number:
Molecular Weight:
99.17
Beilstein/REAXYS Number:
1560282
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

product line

BioReagent

concentration

~40% in 1-butanol

refractive index

n20/D 1.406

suitability

suitable for protein sequencing

storage temp.

−20°C

SMILES string

CN1CCCCC1

InChI

1S/C6H13N/c1-7-5-3-2-4-6-7/h2-6H2,1H3

InChI key

PAMIQIKDUOTOBW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Special packaging for direct use in common sequencers.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3

target_organs

Central nervous system, Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

82.4 °F - closed cup

flash_point_c

28 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nithyanandhan Jayaraj et al.
Langmuir : the ACS journal of surfaces and colloids, 25(24), 13820-13832 (2010-06-22)
Supramolecular complexation behavior of cucurbiturils with paramagnetic nitroxide spin probes was examined by (1)H NMR, X-ray diffraction studies of crystals, computation, and EPR. Both cucurbit[7]uril (CB7) and cucurbit[8]uril (CB8) form a 1:1 complex with 4-(N,N,N-trimethylammonium)-2,2,6,6-tetramethylpiperidinyl-N-oxy bromide (CAT1). The structure of
Bing Yu et al.
Bioorganic & medicinal chemistry letters, 22(1), 110-114 (2011-12-16)
We report herein the design and synthesis of novel 4-aminoquinazoline derivatives based on the inhibitors of VEGFR-2 tyrosine kinases. The VEGFR-2 inhibitory activities of these newly synthesized compounds were also evaluated and compared with that of ZD6474. We found that
A Kolocouris et al.
The Journal of organic chemistry, 66(15), 4989-4997 (2001-07-21)
When a 1-adamantyl or a 2-adamantyl substituent is introduced at the 2-position in N-methylpiperidine, four different chair conformations are possible. Experimental observation using dynamic NMR spectroscopy and molecular mechanics calculations agree that the chair conformation with an equatorial adamantyl group
Shahid Ullah et al.
Journal of chromatography. A, 1218(37), 6388-6395 (2011-07-28)
A trace analytical method based on high performance liquid chromatography coupled to quadrupole time-of-flight high resolution mass spectrometry was developed for simultaneous determination of perfluoroalkyl phosphonates (PFPAs, carbon chain lengths C6,8,10), perfluoroalkyl carboxylates (PFCAs, C5-12), and perfluoroalkyl sulfonates (PFSAs, C4,6,8,10)
[Studies of pyrazine derivatives. XXV. Synthesis and the tuberculostatic activity of the products of reaction of 5-(6-methoxy- and 5-(6 morpholinopyrazinyl)-1,3,4-oxadiazol-2-thione with amines].
D Pancechowska-Ksepko et al.
Acta poloniae pharmaceutica, 45(5), 373-379 (1988-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service