Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

534021

Sigma-Aldrich

2-Propanol

99%, histological grade

Synonym(s):

sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOH
CAS Number:
Molecular Weight:
60.10
Beilstein/REAXYS Number:
635639
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
assay:
99%
application(s):
histology
bp:
82 °C (lit.)
vapor pressure:
33 mmHg ( 20 °C)
44 mmHg ( 25 °C)

Product Name

2-Propanol, histological grade, 99%

vapor density

2.1 (vs air)

vapor pressure

33 mmHg ( 20 °C)
44 mmHg ( 25 °C)

assay

99%

autoignition temp.

750 °F

expl. lim.

2.0-12.7 %, 93 °C

impurities

≤0.5% water

evapn. residue

<0.001%

refractive index

n20/D 1.377 (lit.)

bp

82 °C (lit.)

mp

−89.5 °C (lit.)

density

0.785 g/mL at 25 °C (lit.)

application(s)

histology

SMILES string

CC(C)O

InChI

1S/C3H8O/c1-3(2)4/h3-4H,1-2H3

InChI key

KFZMGEQAYNKOFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

53.6 °F - closed cup

flash_point_c

12.0 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas W Winkler et al.
PLoS genetics, 11(10), e1005378-e1005378 (2015-10-02)
Genome-wide association studies (GWAS) have identified more than 100 genetic variants contributing to BMI, a measure of body size, or waist-to-hip ratio (adjusted for BMI, WHRadjBMI), a measure of body shape. Body size and shape change as people grow older
M Lindberg et al.
Acta dermato-venereologica, 71(5), 384-388 (1991-01-01)
Normal human skin was exposed to two different detergents, sodium lauryl sulphate in distilled water and non-anoic acid in isopropanol at different concentrations. The detergents were applied under occlusion in epicutaneous tests for 24 h and biopsies were taken at
Swapna Ganapathy et al.
Journal of the American Chemical Society, 136(46), 16335-16344 (2014-10-24)
Fundamental research into the Li-O2 battery system has gone into high gear, gaining momentum because of its very high theoretical specific energy. Much progress has been made toward understanding the discharge mechanism, but the mechanism of the oxygen evolution reaction
Emilie Montastier et al.
PLoS computational biology, 11(1), e1004047-e1004047 (2015-01-16)
Nutrigenomics investigates relationships between nutrients and all genome-encoded molecular entities. This holistic approach requires systems biology to scrutinize the effects of diet on tissue biology. To decipher the adipose tissue (AT) response to diet induced weight changes we focused on
Wei-Yu Lin et al.
Human molecular genetics, 24(1), 285-298 (2014-08-30)
Previous studies have suggested that polymorphisms in CASP8 on chromosome 2 are associated with breast cancer risk. To clarify the role of CASP8 in breast cancer susceptibility, we carried out dense genotyping of this region in the Breast Cancer Association

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service