Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

53295

Supelco

Octadecanoic acid tryptamide

Synonym(s):

N-Octadecanoyltryptamin, N-Stearoyltryptamin, N-[2-(3-Indolyl)ethyl]octadecanamide, SAT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H46N2O
CAS Number:
Molecular Weight:
426.68
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

functional group

amide
carboxylic acid

SMILES string

CCCCCCCCCCCCCCCCCC(=O)NCCc1c[nH]c2ccccc12

InChI

1S/C28H46N2O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-21-28(31)29-23-22-25-24-30-27-20-18-17-19-26(25)27/h17-20,24,30H,2-16,21-23H2,1H3,(H,29,31)

InChI key

AIIVCXFBTONHDU-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luana A Biondo et al.
Frontiers in pharmacology, 9, 452-452 (2018-06-06)
Doxorubicin (DX) is a chemotherapeutic drug that is used in clinical practice that promotes deleterious side effects in non-tumor tissues such as adipose tissue. We showed that DX leads to extensive damage in adipose tissue via a disruption in 5'-adenosine
Shinji Matsumura et al.
Nature communications, 10(1), 3441-3441 (2019-08-03)
Eurasian continent has experienced cold winters over the past two decades in contrast with Arctic warming. Previous studies have suggested that the cold Eurasian winters are associated with Arctic sea-ice loss, while others attributed them to atmospheric internal variability. However
Gil Yosef et al.
Scientific reports, 8(1), 996-996 (2018-01-19)
Afforestation is an important approach to mitigate global warming. Its complex interactions with the climate system, however, makes it controversial. Afforestation is expected to be effective in the tropics where biogeochemical and biogeophysical effects act in concert; however, its potential
K H Pietiläinen et al.
International journal of obesity (2005), 40(4), 654-661 (2015-10-27)
Little is known about epigenetic alterations associated with subcutaneous adipose tissue (SAT) in obesity. Our aim was to study genome-wide DNA methylation and gene expression differences in SAT in monozygotic (MZ) twin pairs who are discordant for body mass index
Matteo Buti et al.
GigaScience, 7(4), 1-14 (2018-04-17)
The genus Potentilla is closely related to that of Fragaria, the economically important strawberry genus. Potentilla micrantha is a species that does not develop berries but shares numerous morphological and ecological characteristics with Fragaria vesca. These similarities make P. micrantha

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service