Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

52940

Sigma-Aldrich

1-Hexene

purum, ≥96.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH=CH2
CAS Number:
Molecular Weight:
84.16
Beilstein/REAXYS Number:
1209240
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

3 (vs air)

vapor pressure

155 mmHg ( 21.1 °C)

grade

purum

assay

≥96.0% (GC)

autoignition temp.

487 °F

refractive index

n20/D 1.388 (lit.)
n20/D 1.388

bp

60-66 °C (lit.)

density

0.678 g/mL at 25 °C (lit.)

SMILES string

CCCCC=C

InChI

1S/C6H12/c1-3-5-6-4-2/h3H,1,4-6H2,2H3

InChI key

LIKMAJRDDDTEIG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

pictograms

FlameHealth hazard

signalword

Danger

hcodes

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 2

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

-13.0 °F - closed cup

flash_point_c

-25.0 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Astrid-Caroline Knall et al.
Tetrahedron letters, 55(34), 4763-4766 (2014-08-26)
Inverse electron demand Diels-Alder additions (iEDDA) between 1,2,4,5-tetrazines and olefins have recently found widespread application as a novel 'click chemistry' scheme and as a mild technique for the modification of materials. Norbornenes are, due to their straightforward synthetic availability, especially

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service